Assay Details
Gene Symbol: AES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299736.2 | XP_015155222.1 | 1 - 2 | 227 | 78 | |
XM_015299737.2 | XP_015155223.1 | 1 - 2 | 231 | 78 | ||
GenBank mRNA | BX933376.2 | - | 1 - 2 | 186 | 78 | |
CR338750.1 | - | 1 - 2 | 87 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015299736.2, XM_015299737.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
