Assay Details
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938120.3 | XP_004938177.1 | 3 - 4 | 3626 | 83 | |
XM_015293875.2 | XP_015149361.1 | 3 - 4 | 589 | 83 | ||
XM_015293881.2 | XP_015149367.1 | 3 - 4 | 2970 | 83 | ||
GenBank mRNA | BX933818.1 | - | 2 - 3 | 338 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
