Assay Details
Gene Symbol: C15H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015275416.2 | XP_015130902.1 | 5 - 6 | 73 | ||
XM_025155730.1 | XP_025011498.1 | 5 - 6 | 73 | |||
GenBank mRNA | BX934178.1 | - | 3 - 4 | 541 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
