Assay Details
Gene Symbol: PPIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231332.5 | XP_001231333.3 | 3 - 4 | 1936 | 73 | |
GenBank mRNA | BX935257.1 | - | 3 - 4 | 317 | 73 | |
CR352486.1 | - | 3 - 4 | 388 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
