Assay Details
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294583.2 | XP_015150069.1 | 1 - 2 | 371 | 118 | |
GenBank mRNA | BX935726.1 | - | 1 - 2 | 348 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
