Assay Details
Gene Symbol: LOC100859848 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641852.4 | XP_003641900.1 | 3 - 4 | 1745 | 108 | |
GenBank mRNA | BX950335.2 | - | 2 - 3 | 166 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
