Assay Details
Gene Symbol: NDUFB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015274582.2 | XP_015130068.1 | 1 - 2 | 164 | 116 | |
GenBank mRNA | BX950780.2 | - | 1 - 2 | 140 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
