Assay Details
Gene Symbol: ADRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293684.2 | XP_015149170.1 | 2 | 3863 | 67 | |
GenBank mRNA | CR406855.1 | - | 1 | 924 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
