Assay Details
Gene Symbol: ARTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291307.2 | XP_015146793.1 | 1 - 2 | 82 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015291307.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
