Assay Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015285189.2 | XP_015140675.1 | 5 - 6 | 2513 | 84 | |
XM_015285190.2 | XP_015140676.1 | 5 - 6 | 873 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
