Assay Details
Gene Symbol: MYO15L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295329.2 | XP_015150815.1 | 34 - 35 | 4139 | 111 | |
XM_015295330.2 | XP_015150816.1 | 34 - 35 | 4097 | 111 | ||
XM_025141801.1 | XP_024997569.1 | 32 - 33 | 4443 | 111 | ||
XM_025141802.1 | XP_024997570.1 | 32 - 33 | 4440 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
