Assay Details
Gene Symbol: LOC776719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004948400.3 | XP_004948457.1 | 14 - 15 | 2991 | 73 | |
XM_025143435.1 | XP_024999203.1 | 14 - 15 | 3013 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
