Assay Details
Gene Symbol: HAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294381.2 | XP_015149867.2 | 3 - 4 | 388 | 61 | |
XM_015294390.2 | XP_015149876.2 | 3 - 4 | 388 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
