Assay Details
Gene Symbol: GAS2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004946505.3 | XP_004946562.1 | 5 - 6 | 2100 | 56 | |
XM_425395.6 | XP_425395.4 | 6 - 7 | 2026 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
