Assay Details
Gene Symbol: DCSTAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025148096.1 | XP_025003864.1 | 3 - 4 | 1177 | 154 | |
XM_418376.6 | XP_418376.3 | 2 - 3 | 1107 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() |
