Assay Details
Gene Symbol: COL2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279683.2 | XP_015135169.1 | 46 - 47 | 3692 | 65 | |
XM_025156090.1 | XP_025011858.1 | 46 - 47 | 3692 | 65 | ||
XM_025156091.1 | XP_025011859.1 | 46 - 47 | 3689 | 65 | ||
XM_025156092.1 | XP_025011860.1 | 45 - 46 | 3638 | 65 | ||
XM_025156093.1 | XP_025011861.1 | 45 - 46 | 3638 | 65 | ||
XM_025156094.1 | XP_025011862.1 | 44 - 45 | 3527 | 65 | ||
XM_025156095.1 | XP_025011863.1 | 46 - 47 | 3692 | 65 | ||
XM_025156096.1 | XP_025011864.1 | 46 - 47 | 3898 | 65 | ||
XM_025156097.1 | XP_025011865.1 | 42 - 43 | 3549 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
