Assay Details
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234206.5 | XP_001234207.4 | 2 - 3 | 2734 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234206.5 | XP_001234207.4 | 2 - 3 | 2734 | 85 |
Set Membership: |
![]() ![]() ![]() |