Assay Details
Gene Symbol: LOC101749023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015278810.2 | XP_015134296.2 | 6 - 7 | 1401 | 98 | |
XM_015278817.2 | XP_015134303.2 | 6 - 7 | 1467 | 98 | ||
XM_015278822.2 | XP_015134308.2 | 6 - 7 | 2108 | 98 | ||
XM_015278832.2 | XP_015134318.2 | 5 - 6 | 1243 | 98 | ||
XM_015278845.2 | XP_015134331.2 | 3 - 4 | 971 | 98 | ||
XM_015278851.2 | XP_015134337.2 | 4 - 5 | 1073 | 98 | ||
XM_015278861.2 | XP_015134347.2 | 3 - 4 | 884 | 98 | ||
XM_015278875.2 | XP_015134361.2 | 3 - 4 | 896 | 98 | ||
XM_025146839.1 | XP_025002607.1 | 5 - 6 | 1231 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
