Assay Details
Gene Symbol: GRASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015300270.2 | XP_015155756.2 | 4 - 5 | 479 | 112 | |
XM_025145553.1 | XP_025001321.1 | 4 - 5 | 555 | 112 | ||
XM_025145554.1 | XP_025001322.1 | 3 - 4 | 409 | 112 | ||
XM_025145555.1 | XP_025001323.1 | 2 - 3 | 482 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
