Assay Details
Gene Symbol: FAM46D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420263.6 | XP_420263.3 | 2 - 3 | 247 | 155 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_420263.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
