Assay Details
Gene Symbol: LLGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294417.2 | XP_015149903.2 | 9 - 10 | 5139 | 162 | |
XM_015294418.2 | XP_015149904.2 | 9 - 10 | 5139 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
