Assay Details
Gene Symbol: IL9R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294508.2 | XP_015149994.1 | 8 - 9 | 1062 | 108 | |
XM_015294509.2 | XP_015149995.1 | 7 - 8 | 968 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
