Assay Details
Gene Symbol: PDZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298209.2 | XP_015153695.1 | 12 - 13 | 2559 | 73 | |
XM_025143256.1 | XP_024999024.1 | 9 - 10 | 1330 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
