Assay Details
Gene Symbol: DUSP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294932.2 | XP_015150418.1 | 1 - 2 | 784 | 105 | |
XM_025155556.1 | XP_025011324.1 | 2 - 3 | 558 | 105 | ||
XM_025155557.1 | XP_025011325.1 | 2 - 3 | 492 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
