Assay Details
Gene Symbol: RHOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936113.3 | XP_004936170.1 | 2 - 3 | 371 | 114 | |
XM_025150080.1 | XP_025005848.1 | 2 - 3 | 377 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004936113.3, XM_025150080.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
