Assay Details
Gene Symbol: CCDC160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298185.2 | XP_015153671.1 | 13 - 14 | 1522 | 67 | |
XM_015298186.2 | XP_015153672.1 | 13 - 14 | 1502 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
