Assay Details
Gene Symbol: SEOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252909.1 | NP_001239838.1 | 5 - 6 | 1014 | 108 | |
GenBank mRNA | HM162869.1 | - | 5 - 6 | 1014 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
