Assay Details
Gene Symbol: SEOI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253296.1 | NP_001240225.1 | 2 - 3 | 231 | 96 | |
GenBank mRNA | HM162866.1 | - | 2 - 3 | 231 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
