Assay Details
Gene Symbol: SEOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254039.1 | NP_001240968.1 | 1 - 2 | 191 | 70 | |
GenBank mRNA | HM162859.1 | - | 1 - 2 | 191 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
