Assay Details
Gene Symbol: SEOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254295.1 | NP_001241224.1 | 5 - 6 | 980 | 101 | |
GenBank mRNA | HM162873.1 | - | 5 - 6 | 980 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
