Assay Details
Gene Symbol: SEOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255179.1 | NP_001242108.1 | 5 - 6 | 964 | 124 | |
GenBank mRNA | HM162870.1 | - | 5 - 6 | 964 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
