Assay Details
Gene Symbol: SEOL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255625.1 | NP_001242554.1 | 3 - 4 | 712 | 114 | |
GenBank mRNA | HM162868.1 | - | 3 - 4 | 712 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
