Assay Details
Gene Symbol: ZEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289332.1 | NP_001276261.1 | 12 - 13 | 1707 | 108 | |
GenBank mRNA | AK287187.1 | - | 12 - 13 | 1781 | 108 | |
HM366149.1 | - | 12 - 13 | 1707 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
