Assay Details
Gene Symbol: ZEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289332.1 | NP_001276261.1 | 15 - 16 | 1930 | 80 | |
GenBank mRNA | AK287187.1 | - | 15 - 16 | 2004 | 80 | |
HM366149.1 | - | 15 - 16 | 1930 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
