Assay Details
Gene Symbol: AOC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253104.2 | NP_001240033.1 | 1 | 117 | 117 | |
GenBank mRNA | HM803111.1 | - | 1 | 117 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
