Assay Details
Gene Symbol: AOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255797.1 | NP_001242726.1 | 1 | 118 | 83 | |
GenBank mRNA | HM803110.1 | - | 1 | 118 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the HM803110.1, NM_001255797.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
