Assay Details
Gene Symbol: COL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001292012.1 | NP_001278941.1 | 1 | 515 | 86 | |
GenBank mRNA | HM068898.1 | - | 1 | 515 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
