Assay Details
Gene Symbol: COL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001292014.1 | NP_001278943.1 | 1 | 420 | 64 | |
GenBank mRNA | AK244779.1 | - | 1 | 417 | 64 | |
HM068896.1 | - | 1 | 425 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
