Assay Details
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242.4 | NP_001233.1 | 2 - 3 | 476 | 76 | |
XM_017020232.1 | XP_016875721.1 | 2 - 3 | 494 | 76 | ||
XM_017020234.1 | XP_016875723.1 | 2 - 3 | 494 | 76 | ||
GenBank mRNA | AK315732.1 | - | 2 - 3 | 495 | 76 | |
BC012160.1 | - | 2 - 3 | 462 | 4575359 | 76 | |
M63928.1 | - | 2 - 3 | 365 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
