Assay Details
Gene Symbol: PGM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172818.1 | NP_001166289.1 | 6 - 7 | 1332 | 123 | |
NM_001172819.1 | NP_001166290.1 | 6 - 7 | 1123 | 123 | ||
NM_002633.2 | NP_002624.2 | 6 - 7 | 1246 | 123 | ||
GenBank mRNA | AK293298.1 | - | 6 - 7 | 1123 | 123 | |
AK294186.1 | - | 7 - 8 | 1205 | 123 | ||
AK298505.1 | - | 6 - 7 | 1053 | 123 | ||
AK300100.1 | - | 6 - 7 | 1318 | 123 | ||
AK312254.1 | - | 6 - 7 | 1111 | 123 | ||
BC001756.2 | - | 6 - 7 | 1159 | 3505510 | 123 | |
BC019920.1 | - | 6 - 7 | 1246 | 5122261 | 123 | |
BC067763.1 | - | 6 - 7 | 1124 | 5286530 | 123 | |
BC090856.1 | - | 6 - 7 | 1091 | 123 | ||
BT006961.1 | - | 6 - 7 | 1033 | 123 | ||
M83088.1 | - | 6 - 7 | 1095 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000722.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | ENST00000371084.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | ENST00000538673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | ENST00000540265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01011468.hg.1 | TC01000722.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | TC0100008591.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160947.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 155063 | - | - |
