Assay Details
Gene Symbol: CHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB209329.1 | - | 23 - 24 | 3510 | 110 | |
AF002246.1 | - | 24 - 25 | 3362 | 110 | ||
AK126878.1 | - | 23 - 24 | 3664 | 110 | ||
BC104918.1 | - | 21 - 22 | 3074 | 8143921 | 110 | |
BC143496.1 | - | 21 - 22 | 3071 | 9052004 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
