Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008704.hg.1 |
TC05000628.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008704.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008704.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159100.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159100.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159100.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159100.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159100.hg.1 |
TC0500008619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
ENST00000296870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008619.hg.1 |
NM_000588.3 |
Refseq |