Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000331320.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000405646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000406191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000426567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000434050.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000435036.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000438610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000481206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000481635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000494981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000552286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
NM_001203258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
NM_004689.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
TC1400010703.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000426567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000331320.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000405646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000406191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000434050.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000435036.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000438610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000481206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000481635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000494981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
ENST00000552286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
NM_001203258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181110.hg.1 |
NM_004689.3 |
Refseq |