Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014415.hg.1 |
ENST00000284292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014415.hg.1 |
ENST00000412681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014415.hg.1 |
ENST00000526916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014415.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014415.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014415.hg.1 |
TC11001143.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014414.hg.1 |
ENST00000284292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014414.hg.1 |
ENST00000412681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014414.hg.1 |
ENST00000526916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014414.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014414.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014414.hg.1 |
TC11001143.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014416.hg.1 |
ENST00000284292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014416.hg.1 |
ENST00000412681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014416.hg.1 |
ENST00000526916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014416.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014416.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014416.hg.1 |
TC11001143.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
TC1100009398.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170387.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
TC1100009398.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170389.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
ENST00000412681.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009398.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170386.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170386.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170386.hg.1 |
NM_006176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170386.hg.1 |
TC1100009398.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100170386.hg.1 |
ENST00000284292.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170386.hg.1 |
NM_001126181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170386.hg.1 |
NM_006176.2 |
Refseq |