Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015578.hg.1 |
ENST00000224605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015578.hg.1 |
ENST00000374247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015578.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015578.hg.1 |
TC10001252.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015580.hg.1 |
ENST00000224605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015580.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015580.hg.1 |
TC10001252.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000151333.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151333.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151333.hg.1 |
TC1000007543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000151333.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151333.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151332.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151332.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151332.hg.1 |
TC1000007543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000151332.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151332.hg.1 |
NM_004962.4 |
Refseq |