Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008674.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008674.hg.1 |
ENST00000412089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008674.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008674.hg.1 |
TC03000417.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008680.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008680.hg.1 |
ENST00000412089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008680.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008680.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008680.hg.1 |
TC03000417.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008684.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008684.hg.1 |
ENST00000412089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008684.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008684.hg.1 |
TC03000417.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008686.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008686.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008686.hg.1 |
TC03000417.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
TC0300007745.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155953.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
TC0300007745.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155957.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
TC0300007745.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155958.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155946.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155946.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155946.hg.1 |
TC0300007745.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155946.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155946.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000273258.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000470936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000478935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000484921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
ENST00000485444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007745.hg.1 |
NM_006407.3 |
Refseq |