Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005790.hg.1 |
ENST00000381020.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005790.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005790.hg.1 |
TC13000513.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005789.hg.1 |
ENST00000381020.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005789.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005789.hg.1 |
TC13000513.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005791.hg.1 |
ENST00000381020.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005791.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005791.hg.1 |
TC13000513.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008449.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156951.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156951.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156951.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156951.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156951.hg.1 |
TC1300008449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156947.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156947.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156947.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156947.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156947.hg.1 |
ENST00000548877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156947.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156947.hg.1 |
TC1300008449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156949.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156949.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156949.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156949.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156949.hg.1 |
TC1300008449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156950.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156950.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156950.hg.1 |
ENST00000381020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156950.hg.1 |
NM_001265.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156950.hg.1 |
TC1300008449.hg.1 |
Clariom D TC |