Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027419.hg.1 |
TC05001970.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027419.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027419.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027421.hg.1 |
TC05001970.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027421.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027421.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027420.hg.1 |
TC05001970.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027420.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027420.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187647.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187647.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187647.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187647.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187647.hg.1 |
TC0500012553.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500187648.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187648.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187648.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187648.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187648.hg.1 |
TC0500012553.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500187651.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187651.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187651.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187651.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187651.hg.1 |
TC0500012553.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500187650.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187650.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187650.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187650.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187650.hg.1 |
TC0500012553.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500187649.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187649.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187649.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187649.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187649.hg.1 |
TC0500012553.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
ENST00000231121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012553.hg.1 |
NM_004821.2 |
Refseq |