Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001098.hg.1 |
TC05000096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001098.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001098.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001095.hg.1 |
TC05000096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001095.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001095.hg.1 |
ENST00000447228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001095.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001099.hg.1 |
TC05000096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001099.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001099.hg.1 |
ENST00000447228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001099.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001100.hg.1 |
TC05000096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001100.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001100.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001102.hg.1 |
TC05000096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001102.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001102.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001097.hg.1 |
TC05000096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001097.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001097.hg.1 |
ENST00000447228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001097.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192765.hg.1 |
TC0500013155.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192762.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192762.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192762.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192762.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192762.hg.1 |
TC0500013155.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192764.hg.1 |
TC0500013155.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500192761.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192761.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192761.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192761.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192761.hg.1 |
TC0500013155.hg.1 |
Clariom D TC |