Assay Details
Gene Symbol: NIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011536823.2 | XP_011535125.1 | 1 - 2 | 154 | 60 | |
GenBank mRNA | AF212162.2 | - | 1 - 2 | 332 | 60 |
Gene Symbol: LOC105370489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135258.1 | 1 - 2 | 60 | |||
GenBank mRNA | HY010266.1 | - | 1 - 2 | 60 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
